Assay Details
Target Gene Details
Entrez Gene ID: | 400746 |
Gene Name: | non-compact myelin associated protein |
Gene Aliases: |
C1orf130, MP11 |
Location: |
Chr.1:24556076-24609328 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCMAP | NM_001010980.4 | NP_001010980.1 | ||
XM_011541463.2 | XP_011539765.1 | |||
AK124519.1 | ||||
CB961010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545823 | Chr.1:24568506 - 24621457 on Build GRCh38 | Gain | NCMAP |
dgv31e199 | Chr.1:24569169 - 24574377 on Build GRCh38 | Deletion | NCMAP |
nsv436645 | Chr.1:24568967 - 24575752 on Build GRCh38 | Deletion | NCMAP |
esv27046 | Chr.1:24569305 - 24574653 on Build GRCh38 | Loss | NCMAP |
nsv1147629 | Chr.1:24569262 - 24574370 on Build GRCh38 | Deletion | NCMAP |
esv3585506 | Chr.1:24569283 - 24574380 on Build GRCh38 | Loss | NCMAP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |