Assay Details
Target Gene Details
Entrez Gene ID: | 10487 |
Gene Name: | adenylate cyclase associated protein 1 |
Gene Aliases: |
CAP, CAP1-PEN |
Location: |
Chr.1:40040070-40072649 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss | PPT1 LOC101929536 TRIT1 OXCT2 BMP8B RLF MYCL CAP1 PPIE MFSD2A |
nsv950665 | Chr.1:40031829 - 40062428 on Build GRCh38 | Duplication | CAP1 |
esv2750836 | Chr.1:40038107 - 40093576 on Build GRCh38 | Gain | PPT1 CAP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |