Assay Details
Target Gene Details
Entrez Gene ID: | 56267 |
Gene Name: | kynurenine aminotransferase 3 |
Gene Aliases: |
CCBL2, KAT3, KATIII |
Location: |
Chr.1:88935773-88992960 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KYAT3 | NM_001008661.2 | NP_001008661.1 | ||
NM_001008662.2 | NP_001008662.1 | |||
AK057176.1 | ||||
AK301326.1 | ||||
AK315917.1 | ||||
AL833593.1 | ||||
AY028624.1 | AAK26163.1 | |||
BM728982.1 | ||||
CR627392.1 | CAH10487.1 | |||
DC424120.1 |
Target Gene Details
Entrez Gene ID: | 494115 |
Gene Name: | RNA binding motif protein, X-linked like 1 |
Gene Aliases: |
RBM1 |
Location: |
Chr.1:88979456-88992960 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMXL1 | NM_001162536.2 | NP_001156008.1 | ||
NM_019610.5 | NP_062556.2 | |||
AL832554.1 | CAI46148.1 | |||
BC012942.1 | AAH12942.1 | |||
DA010727.1 | ||||
DA216732.1 | ||||
DA390224.1 | ||||
DC397531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv207n100 | Chr.1:88932165 - 88995971 on Build GRCh38 | Loss | KYAT3 RBMXL1 |
esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain | KYAT3 GBP1 GBP3 RBMXL1 |
nsv1002798 | Chr.1:88954378 - 88988906 on Build GRCh38 | Gain | KYAT3 RBMXL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |