Assay Details
Target Gene Details
Entrez Gene ID: | 100287898 |
Gene Name: | tetratricopeptide repeat domain 34 |
Gene Aliases: |
- |
Location: |
Chr.1:2636988-2789737 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC34 | NM_001242672.1 | NP_001229601.1 | ||
XM_016999990.1 | XP_016855479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585032 | Chr.1:2776470 - 2780643 on Build GRCh38 | Loss | TTC34 |
esv3585031 | Chr.1:2767758 - 2802942 on Build GRCh38 | Gain | TTC34 |
esv3583359 | Chr.1:2777970 - 3286320 on Build GRCh38 | Gain | TTC34 LINC00982 PRDM16 ACTRT2 MIR4251 |
esv2661153 | Chr.1:2776483 - 2780558 on Build GRCh38 | Deletion | TTC34 |
dgv11n100 | Chr.1:2777886 - 3350773 on Build GRCh38 | Gain | TTC34 LINC00982 LOC105378604 PRDM16 ACTRT2 MIR4251 |
nsv428399 | Chr.1:2622539 - 3235926 on Build GRCh38 | Gain | TTC34 MMEL1 LINC00982 PRDM16 ACTRT2 MIR4251 |
nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss | TTC34 MMEL1 PLCH2 PANK4 TNFRSF14 PEX10 LOC115110 FAM213B RER1 LOC100996583 HES5 |
esv23086 | Chr.1:2767708 - 2778413 on Build GRCh38 | Gain+Loss | TTC34 |