Assay Details
Target Gene Details
Entrez Gene ID: | 51060 |
Gene Name: | thioredoxin domain containing 12 |
Gene Aliases: |
AG1, AGR1, ERP16, ERP18, ERP19, PDIA16, TLP19, hAG-1, hTLP19 |
Location: |
Chr.1:52020131-52056171 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNDC12 | NM_015913.3 | NP_056997.1 | ||
AF131758.1 | AAD20035.1 | |||
AF543416.1 | AAN34781.1 | |||
AK056169.1 | ||||
AK075409.1 | ||||
AY358982.1 | AAQ89341.1 | |||
BC001493.2 | AAH01493.1 | |||
BC008913.2 | AAH08913.1 | |||
BC008953.2 | AAH08953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829848 | Chr.1:51902939 - 52076027 on Build GRCh38 | Loss | BTF3L4 TXNDC12 TXNDC12-AS1 KTI12 RAB3B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |