Assay Details
Target Gene Details
Entrez Gene ID: | 100131998 |
Gene Name: | RRN3 homolog, RNA polymerase I transcription factor pseudogene 3 |
Gene Aliases: |
- |
Location: |
Chr.16:22419546-22437715 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RRN3P3 | NR_027460.1 | |||
AK055742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052004 | Chr.16:22406984 - 22693199 on Build GRCh38 | Gain | NPIPB5 RRN3P3 MFSD13B SMG1P1 LOC653786 |
esv3638207 | Chr.16:22418547 - 22435682 on Build GRCh38 | Gain | RRN3P3 |
nsv1160373 | Chr.16:22420049 - 22546990 on Build GRCh38 | Duplication | NPIPB5 RRN3P3 SMG1P1 LOC653786 |
dgv2813n100 | Chr.16:22412835 - 22699409 on Build GRCh38 | Loss | NPIPB5 RRN3P3 SMG1P1 LOC653786 |
nsv428324 | Chr.16:22419254 - 22760988 on Build GRCh38 | Gain | NPIPB5 RRN3P3 SMG1P1 LOC653786 |
nsv571645 | Chr.16:22379935 - 22438562 on Build GRCh38 | Loss | RRN3P3 MFSD13B SMG1P1 |
esv2758637 | Chr.16:22282803 - 22760988 on Build GRCh38 | Gain+Loss | NPIPB5 RRN3P3 MFSD13B POLR3E CDR2 SMG1P1 LOC653786 LOC440346 EEF2K |