Assay Details
Target Gene Details
Entrez Gene ID: | 123803 |
Gene Name: | N-terminal asparagine amidase |
Gene Aliases: |
PNAA, PNAD |
Location: |
Chr.16:15037853-15056079 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTAN1 | NM_001270766.1 | NP_001257695.1 | ||
NM_001270767.1 | NP_001257696.1 | |||
NM_173474.3 | NP_775745.1 | |||
XM_011522355.2 | XP_011520657.1 | |||
AF092440.1 | AAP97215.1 | |||
AK092915.1 | ||||
BC017336.2 | AAH17336.1 |
Target Gene Details
Entrez Gene ID: | 23042 |
Gene Name: | pyridoxal dependent decarboxylase domain containing 1 |
Gene Aliases: |
LP8165 |
Location: |
Chr.16:14974591-15139410 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDXDC1 | NM_001285444.1 | 22 | 4156 | NP_001272373.1 |
NM_001285445.1 | 22 | 4153 | NP_001272374.1 | |
NM_001285447.1 | 23 | 4049 | NP_001272376.1 | |
NM_001285448.1 | 22 | 4062 | NP_001272377.1 | |
NM_001285449.1 | NP_001272378.1 | |||
NM_001324019.1 | 23 | 4234 | NP_001310948.1 | |
NM_001324020.1 | NP_001310949.1 | |||
NM_001324021.1 | NP_001310950.1 | |||
NM_015027.3 | 23 | 4237 | NP_055842.2 | |
XM_017023065.1 | XP_016878554.1 | |||
AY203955.1 | AAP34478.1 | |||
BC045554.1 | AAH45554.2 | |||
BC060871.1 | 23 | 4071 | AAH60871.1 | |
BM997114.1 | 1 | 437 | ||
CD370048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638022 | Chr.16:15031773 - 15047500 on Build GRCh38 | Loss |
![]() ![]() |
dgv4919n54 | Chr.16:14939348 - 15040518 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3570354 | Chr.16:15034129 - 15223463 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1042130 | Chr.16:14954899 - 15100193 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv509598 | Chr.16:15025400 - 15067337 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv1038955 | Chr.16:14885100 - 15322641 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2762254 | Chr.16:14741210 - 15322653 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv2755n100 | Chr.16:14877546 - 15434384 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3892796 | Chr.16:14961008 - 15074903 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3638020 | Chr.16:15031082 - 15082591 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv4922n54 | Chr.16:15025750 - 15099845 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2758632 | Chr.16:14528198 - 15272693 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1053467 | Chr.16:15031584 - 15322641 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)