Assay Details
Target Gene Details
Entrez Gene ID: | 6527 |
Gene Name: | solute carrier family 5 member 4 |
Gene Aliases: |
DJ90G24.4, SAAT1, SGLT3 |
Location: |
Chr.22:32218423-32355057 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC5A4 | NM_014227.2 | 8 | 785 | NP_055042.1 |
XM_006724308.3 | 9 | 1036 | XP_006724371.1 | |
XM_011530342.2 | 10 | 1154 | XP_011528644.1 | |
XM_011530343.2 | 8 | 700 | XP_011528645.1 | |
XM_011530344.2 | 7 | 665 | XP_011528646.1 | |
XM_017028920.1 | 10 | 1115 | XP_016884409.1 | |
AJ133127.1 | 8 | 799 | CAB81772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3647603 | Chr.22:32234853 - 32235767 on Build GRCh38 | Loss |
![]() |
nsv459874 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3893475 | Chr.22:32228077 - 32260035 on Build GRCh38 | Gain |
![]() |
nsv834178 | Chr.22:32094462 - 32281534 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2672031 | Chr.22:32234881 - 32235746 on Build GRCh38 | Deletion |
![]() |
nsv3597 | Chr.22:32192437 - 32236737 on Build GRCh38 | Deletion |
![]() ![]() |
esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)