Assay Details
Target Gene Details
Entrez Gene ID: | 6304 |
Gene Name: | SATB homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.3:18345388-18445588 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SATB1 | NM_001131010.3 | 11 | 2258 | NP_001124482.1 |
NM_001195470.2 | 12 | 3856 | NP_001182399.1 | |
NM_001322871.1 | 12 | 2402 | NP_001309800.1 | |
NM_001322872.1 | 12 | 2306 | NP_001309801.1 | |
NM_001322873.1 | 11 | 2306 | NP_001309802.1 | |
NM_001322874.1 | 11 | 2120 | NP_001309803.1 | |
NM_001322875.1 | 11 | 2118 | NP_001309804.1 | |
NM_001322876.1 | 10 | 2071 | NP_001309805.1 | |
NM_002971.5 | 11 | 3760 | NP_002962.1 | |
XM_011533988.2 | 12 | 2316 | XP_011532290.1 | |
XM_011533989.2 | 12 | 2216 | XP_011532291.1 | |
AB209761.1 | 12 | 3128 | BAD92998.1 | |
AK127242.1 | 11 | 3765 | ||
BC001744.1 | 11 | 2197 | AAH01744.1 | |
M97287.1 | 11 | 2239 | AAA60304.1 | |
U92993.1 | 2 | 332 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460455 | Chr.3:18344240 - 18386958 on Build GRCh38 | Loss | SATB1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |