Assay Details
Target Gene Details
Entrez Gene ID: | 64756 |
Gene Name: | ATP synthase mitochondrial F1 complex assembly factor 1 |
Gene Aliases: |
ATP11, ATP11p |
Location: |
Chr.1:46635039-46668427 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATPAF1 | NM_001042546.2 | NP_001036011.2 | ||
NM_001243728.1 | NP_001230657.1 | |||
NM_001256418.1 | NP_001243347.1 | |||
NM_022745.4 | NP_073582.3 | |||
AK026004.1 | BAB15316.1 | |||
AK301874.1 | ||||
AK302090.1 | ||||
AK310338.1 | ||||
BC008498.1 | AAH08498.1 | |||
BC018781.1 | AAH18781.1 | |||
BC071731.1 | AAH71731.1 | |||
BX438342.2 | ||||
DA244814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1007280 | Chr.1:46622444 - 46663916 on Build GRCh38 | Gain | ATPAF1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |