Assay Details
Target Gene Details
Entrez Gene ID: | 83594 |
Gene Name: | nudix hydrolase 12 |
Gene Aliases: |
- |
Location: |
Chr.5:103548855-103562805 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT12 | NM_001300741.1 | 3 | 394 | NP_001287670.1 |
NM_031438.3 | 3 | 448 | NP_113626.1 | |
XM_005272095.1 | 3 | 433 | XP_005272152.1 | |
XM_005272097.3 | 3 | 394 | XP_005272154.1 | |
AK098066.1 | 3 | 417 | ||
AK304010.1 | 3 | 382 | ||
AL136592.1 | 3 | 403 | CAB66527.1 | |
BC041099.1 | 3 | 432 | AAH41099.1 | |
CB131118.1 | 3 | 436 | ||
DB029218.1 | 3 | 450 | ||
DC417976.1 | 3 | 383 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3606077 | Chr.5:103549971 - 103712728 on Build GRCh38 | Gain | NUDT12 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |