Assay Details
Target Gene Details
Entrez Gene ID: | 23640 |
Gene Name: | HSPA (Hsp70) binding protein 1 |
Gene Aliases: |
FES1 |
Location: |
Chr.19:55262222-55280383 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPBP1 | NM_001130106.1 | 3 | 308 | NP_001123578.1 |
NM_001297600.1 | 1 | 223 | NP_001284529.1 | |
NM_012267.4 | 2 | 417 | NP_036399.3 | |
XM_005258700.2 | 2 | 148 | XP_005258757.1 | |
XM_005258701.2 | 2 | 210 | XP_005258758.1 | |
XM_011526691.1 | 3 | 306 | XP_011524993.1 | |
XM_017026543.1 | 2 | 145 | XP_016882032.1 | |
XM_017026544.1 | 2 | 145 | XP_016882033.1 | |
XM_017026545.1 | 2 | 148 | XP_016882034.1 | |
AB020592.1 | 2 | 139 | BAB18742.1 | |
AF093420.1 | 2 | 142 | AAC79703.1 | |
AF187859.1 | 2 | 98 | AAF35833.1 | |
AF217996.1 | 3 | 285 | AAG17238.1 | |
AK075293.1 | 2 | 155 | ||
AK129755.1 | 2 | 211 | ||
AK130781.1 | 2 | 145 | ||
AK294358.1 | 1 | 222 | ||
BC001236.1 | 1 | 63 | AAH01236.1 | |
BC002373.2 | 2 | 129 | AAH02373.1 | |
BM853303.1 | ||||
CR457118.1 | CAG33399.1 | |||
DB500673.1 | 1 | 223 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |