Assay Details
Target Gene Details
Entrez Gene ID: | 113622 |
Gene Name: | ADP-ribosylhydrolase like 1 |
Gene Aliases: |
ARH2 |
Location: |
Chr.13:113421940-113453524 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADPRHL1 | NM_138430.4 | 7 | 1099 | NP_612439.2 |
NM_199162.2 | 7 | 942 | NP_954631.1 | |
AJ313429.1 | 7 | 1099 | CAC86114.1 | |
BC009755.2 | 7 | 942 | AAH09755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain | F7 GRTP1-AS1 PROZ F10 F10-AS1 GRTP1 LOC101928841 MIR8075 TFDP1 PCID2 CUL4A DCUN1D2 LAMP1 LOC105370378 LOC102724474 TMCO3 MCF2L ADPRHL1 |
dgv1744n100 | Chr.13:113384831 - 113442576 on Build GRCh38 | Gain | LOC101928841 ADPRHL1 |
nsv509522 | Chr.13:113408681 - 113432073 on Build GRCh38 | Insertion | LOC101928841 ADPRHL1 |
nsv563318 | Chr.13:113319297 - 113430196 on Build GRCh38 | Loss | GRTP1-AS1 LAMP1 LOC105370378 GRTP1 LOC101928841 ADPRHL1 |
nsv1195 | Chr.13:113393584 - 113427243 on Build GRCh38 | Insertion | LOC101928841 ADPRHL1 |
nsv951928 | Chr.13:113392386 - 113435785 on Build GRCh38 | Deletion | LOC101928841 ADPRHL1 |
nsv563321 | Chr.13:113369086 - 113452383 on Build GRCh38 | Loss | LOC101928841 ADPRHL1 |
esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss | DCUN1D2 LOC101928841 TMCO3 ADPRHL1 |
nsv1044941 | Chr.13:113369130 - 113433835 on Build GRCh38 | Gain | LOC101928841 ADPRHL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |