Assay Details
Target Gene Details
Entrez Gene ID: | 387357 |
Gene Name: | thymocyte selection associated |
Gene Aliases: |
C6orf190, C6orf207, GASP, SPOT, TSEPA |
Location: |
Chr.6:127697191-127918631 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THEMIS | NM_001010923.2 | 6 | 3810 | NP_001010923.1 |
NM_001164685.1 | 7 | 3927 | NP_001158157.1 | |
NM_001164687.1 | 6 | 3774 | NP_001158159.1 | |
NM_001318531.1 | 7 | 3875 | NP_001305460.1 | |
XM_011535813.2 | 8 | 4004 | XP_011534115.1 | |
XM_011535814.2 | 7 | 3775 | XP_011534116.1 | |
XM_011535816.1 | 8 | 4000 | XP_011534118.1 | |
XM_017010849.1 | 6 | 3518 | XP_016866338.1 | |
AK094863.1 | 3 | 2087 | ||
AK128377.1 | 7 | 3808 | ||
BG461112.1 | 1 | 61 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |