Assay Details
Target Gene Details
Entrez Gene ID: | 5650 |
Gene Name: | kallikrein related peptidase 7 |
Gene Aliases: |
PRSS6, SCCE, hK7 |
Location: |
Chr.19:50976479-50984064 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK7 | NM_001207053.1 | 3 | 551 | NP_001193982.1 |
NM_001243126.1 | 3 | 534 | NP_001230055.1 | |
NM_005046.3 | 4 | 682 | NP_005037.1 | |
NM_139277.2 | 4 | 512 | NP_644806.1 | |
AA101043.1 | ||||
AF411214.1 | 4 | 656 | AAN03662.1 | |
AF411215.1 | 3 | 525 | AAN03663.1 | |
AK289660.1 | 4 | 535 | ||
AK295313.1 | 3 | 534 | ||
AY601109.1 | 3 | 410 | ||
AY646152.1 | 2 | 266 | AAT66047.1 | |
BC032005.1 | 4 | 521 | AAH32005.1 | |
DQ018784.1 | 3 | 410 | ||
KF963189.1 | 3 | 374 | ||
KF963190.1 | 3 | 375 | ||
KF963191.1 | 3 | 375 | ||
KF963192.1 | ||||
KF963193.1 | 3 | 375 | ||
L33404.1 | 3 | 425 | AAC37551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064207 | Chr.19:50971269 - 50994022 on Build GRCh38 | Loss | KLK7 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |