Assay Details
Target Gene Details
Entrez Gene ID: | 272 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.11:10450321-10507579 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMPD3 | NM_000480.2 | 2 | 423 | NP_000471.1 |
NM_001025389.1 | 2 | 386 | NP_001020560.1 | |
NM_001025390.1 | 2 | 525 | NP_001020561.1 | |
NM_001172430.1 | 2 | 350 | NP_001165901.1 | |
NM_001172431.1 | 2 | 251 | NP_001165902.1 | |
AK289998.1 | 2 | 157 | ||
AK294444.1 | 2 | 157 | ||
AK295046.1 | 2 | 249 | ||
AK299113.1 | 2 | 98 | ||
AK301507.1 | 2 | 273 | ||
AK302970.1 | 2 | 341 | ||
AK308181.1 | 2 | 193 | ||
AK311436.1 | 3 | 230 | ||
AK311607.1 | 2 | 446 | ||
AK311711.1 | 2 | 410 | ||
AK313039.1 | 2 | 157 | ||
BC126118.1 | 2 | 418 | ||
D12775.1 | 2 | 242 | BAA02240.1 | |
DC334250.1 | 3 | 296 | ||
DC401375.1 | 2 | 350 | ||
M84720.1 | 2 | 257 | AAA58365.1 | |
M84721.1 | 2 | 185 | AAA58366.1 | |
M84722.1 | 2 | 309 | AAA58367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625358 | Chr.11:10435119 - 10512909 on Build GRCh38 | Gain | AMPD3 RNF141 MIR4485 MTRNR2L8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |