Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | 14 | 5205 | NP_001073864.2 |
NM_004920.2 | 13 | 4983 | NP_004911.2 | |
XM_006722193.2 | 14 | 5198 | XP_006722256.1 | |
XM_006722194.2 | 13 | 5050 | XP_006722257.1 | |
XM_006722195.2 | 12 | 4992 | XP_006722258.1 | |
XM_006722196.2 | 14 | 5167 | XP_006722259.1 | |
XM_011525509.2 | 13 | 6634 | XP_011523811.1 | |
XM_011525510.2 | 13 | 5010 | XP_011523812.1 | |
XM_011525511.2 | 9 | 4863 | XP_011523813.1 | |
XM_017025421.1 | 13 | 4997 | XP_016880910.1 | |
XM_017025422.1 | 13 | 5156 | XP_016880911.1 | |
AB014541.2 | 13 | 4983 | ||
AK094931.1 | 6 | 2004 | ||
AK131395.1 | 14 | 4624 | BAD18544.1 | |
AK131529.1 | 16 | 4834 | BAD18667.1 | |
BC047378.1 | 4 | 2770 | AAH47378.1 | |
BI256809.1 | 1 | 188 |
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAIAP2 | NM_001144888.1 | NP_001138360.1 | ||
NM_006340.2 | NP_006331.1 | |||
NM_017451.2 | NP_059345.1 | |||
AB017119.1 | BAA74773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523417 | Chr.17:81115790 - 81164271 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)