Assay Details
Target Gene Details
Entrez Gene ID: | 80821 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
PA-PLA1, PAPLA1, SPG28 |
Location: |
Chr.14:53036740-53153328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDHD1 | NM_001160147.1 | NP_001153619.1 | ||
NM_001160148.1 | NP_001153620.1 | |||
NM_030637.2 | NP_085140.2 | |||
XM_005268102.2 | XP_005268159.1 | |||
XM_005268103.2 | XP_005268160.1 | |||
XM_005268105.2 | XP_005268162.1 | |||
XM_011537188.2 | XP_011535490.1 | |||
XM_011537189.2 | XP_011535491.1 | |||
XM_017021668.1 | XP_016877157.1 | |||
XM_017021669.1 | XP_016877158.1 | |||
AB051492.1 | BAB21796.1 | |||
AK058137.1 | BAB71679.1 | |||
AK125372.1 | ||||
AK125556.1 | ||||
BC018014.1 | AAH18014.1 | |||
BC030703.2 | AAH30703.1 | |||
DQ315474.1 | ABC43196.1 | |||
GU376468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832797 | Chr.14:52932853 - 53113976 on Build GRCh38 | Gain | DDHD1 FERMT2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |