Assay Details
Target Gene Details
Entrez Gene ID: | 5408 |
Gene Name: | pancreatic lipase related protein 2 (gene/pseudogene) |
Gene Aliases: |
PLRP2 |
Location: |
Chr.10:116620953-116645143 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PNLIPRP2 | NM_005396.4 | 4 | 295 | NP_005387.2 |
NR_103727.1 | 4 | 295 | ||
XM_017016339.1 | 3 | 279 | XP_016871828.1 | |
AK291492.1 | 4 | 295 | ||
AK307745.1 | 4 | 294 | ||
AK308508.1 | 4 | 305 | ||
AK308511.1 | 4 | 295 | ||
AK308521.1 | 4 | 305 | ||
BC005989.2 | 3 | 269 | AAH05989.1 | |
BP325072.1 | 4 | 295 | ||
CR456949.1 | 3 | 269 | CAG33230.1 | |
M93284.1 | 3 | 269 | AAA59533.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |