Assay Details
Target Gene Details
Entrez Gene ID: | 1009 |
Gene Name: | cadherin 11 |
Gene Aliases: |
CAD11, CDHOB, OB, OSF-4 |
Location: |
Chr.16:64943753-65126112 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH11 | NM_001308392.1 | 11 | 2159 | NP_001295321.1 |
NM_001797.3 | 11 | 2159 | NP_001788.2 | |
XM_005255761.3 | 11 | 2030 | XP_005255818.1 | |
XM_005255762.2 | 11 | 2244 | XP_005255819.1 | |
XM_005255763.2 | 11 | 1954 | XP_005255820.1 | |
XM_011522803.2 | 10 | 1864 | XP_011521105.1 | |
XM_017022830.1 | 10 | 1719 | XP_016878319.1 | |
AB209760.1 | 9 | 1637 | BAD92997.1 | |
AK291251.1 | 11 | 2085 | ||
AK297377.1 | 11 | 1670 | ||
BC013609.1 | 9 | 1778 | AAH13609.1 | |
D21254.1 | 11 | 2045 | BAA04798.1 | |
D21255.1 | 11 | 2080 | BAA04799.1 | |
L34056.1 | 9 | 1759 | AAA35622.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |