Assay Details
Target Gene Details
Entrez Gene ID: | 146198 |
Gene Name: | ZFP90 zinc finger protein |
Gene Aliases: |
FIK, NK10, ZNF756, zfp-90 |
Location: |
Chr.16:68533484-68576072 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZFP90 | NM_001305203.1 | 5 | 1455 | NP_001292132.1 |
NM_001305204.1 | NP_001292133.1 | |||
NM_001305206.1 | 5 | 1510 | NP_001292135.1 | |
NM_001305207.1 | 5 | 1507 | NP_001292136.1 | |
NM_001305208.1 | 5 | 1500 | NP_001292137.1 | |
NM_133458.3 | 5 | 1445 | NP_597715.2 | |
NR_130976.1 | 5 | 1553 | ||
NR_130977.1 | 5 | 1503 | ||
NR_130978.1 | 5 | 1561 | ||
XM_005255804.2 | 5 | 1385 | XP_005255861.1 | |
XM_017022952.1 | 4 | 1492 | XP_016878441.1 | |
AB075834.1 | 1 | 649 | BAB85540.1 | |
AK056596.1 | 1 | 238 | BAB71228.1 | |
AK074332.1 | 5 | 1469 | ||
AK122847.1 | 1 | 1880 | ||
AK126167.1 | BAC86467.1 | |||
AK304152.1 | 5 | 1347 | ||
AK316251.1 | 5 | 1372 | ||
AW135652.1 | ||||
BC033165.1 | 1 | 142 | ||
BC140886.1 | 5 | 1445 | ||
BC140887.1 | 5 | 1445 | ||
KC249978.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |