Assay Details
Target Gene Details
Entrez Gene ID: | 10158 |
Gene Name: | PDZK1 interacting protein 1 |
Gene Aliases: |
DD96, MAP17, SPAP |
Location: |
Chr.1:47183589-47190099 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDZK1IP1 | NM_005764.3 | 4 | 658 | NP_005755.1 |
BC012303.1 | 4 | 571 | AAH12303.1 | |
BI763294.1 | 2 | 323 | ||
BM924665.1 | 4 | 684 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950709 | Chr.1:47173129 - 47191428 on Build GRCh38 | Deletion | PDZK1IP1 LINC00853 |