Assay Details
Target Gene Details
Entrez Gene ID: | 100131626 |
Gene Name: | uncharacterized LOC100131626 |
Gene Aliases: |
- |
Location: |
Chr.11:118381220-118418815 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100131626 | NR_046369.1 | |||
NR_046370.1 |