Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | 2 | 2527 | NP_057356.1 |
XM_005261315.2 | 2 | 3738 | XP_005261372.1 | |
XM_006724105.3 | 2 | 3044 | XP_006724168.1 | |
XM_017028539.1 | 2 | 4820 | XP_016884028.1 | |
AB051450.1 | 2 | 2286 | ||
BC038957.1 | 2 | 2300 | AAH38957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | TOB2 TEF LOC100996598 RANGAP1 PHF5A ZC3H7B |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | TOB2 TEF ACO2 LOC100996598 RANGAP1 CSDC2 MIR6889 PMM1 PHF5A ZC3H7B LOC105373044 POLR3H |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | TOB2 TEF ACO2 LOC100996598 PHF5A LOC105373044 POLR3H |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | TOB2 TEF LOC100996598 RANGAP1 MIR6889 CHADL ZC3H7B |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | TOB2 MEI1 C22orf46 LOC100996598 RANGAP1 DESI1 CSDC2 L3MBTL2 ZC3H7B XRCC6 EP300 POLR3H SNORD140 TEF ACO2 SNU13 MIR6889 EP300-AS1 PMM1 CHADL PHF5A LOC105373044 MIR1281 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |