Assay Details
Target Gene Details
Entrez Gene ID: | 9091 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Q |
Gene Aliases: |
GPI1, c407A10.1 |
Location: |
Chr.16:569968-584136 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGQ | NM_004204.3 | NP_004195.2 | ||
NM_148920.2 | NP_683721.1 | |||
AB003723.1 | BAA24948.1 | |||
AF030177.1 | AAC32661.1 | |||
AK300297.1 | ||||
AK314361.1 | ||||
BC006377.2 | AAH06377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833113 | Chr.16:506513 - 628160 on Build GRCh38 | Loss | CAPN15 LOC107987233 LOC101929280 RAB40C LINC00235 PIGQ RAB11FIP3 MIR5587 MIR3176 PRR35 NHLRC4 |
esv3637579 | Chr.16:574322 - 585512 on Build GRCh38 | Loss | PIGQ |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | HAGH ERVK13-1 AMDHD2 WFIKKN1 HBM LOC100287175 TELO2 LOC729652 STUB1 LOC106660606 LOC105371049 SSTR5 MIR940 CCDC78 ECI1 RPS2 MEIOB NME3 FAM173A ARHGDIG KCTD5 NTN3 HBZ LOC101929280 SLC9A3R2 WDR24 PTX4 GFER MRPS34 NHLRC4 CHTF18 TPSAB1 RPL23AP5 SPSB3 TPSB2 RGS11 LOC100134368 GNG13 MSLN HBA2 MIR3177 MIR6511B1 CCNF NUBP2 WDR90 DNASE1L2 BRICD5 EME2 IFT140 LUC7L FAM234A HAGHL E4F1 TSR3 PIGQ C1QTNF8 SNORA64 TPSG1 RNPS1 POLR3K HBQ1 MIR6768 UNKL NOXO1 RPL3L RHOT2 CRAMP1 SNORA10 NPW WASIR2 TRAF7 DECR2 MIR3677 LOC107984876 LINC00254 SOX8 MPG RHBDF1 JMJD8 PRR25 MIR3176 ABCA3 CASKIN1 AXIN1 MIR1225 RAB26 NME4 TSC2 PGP TMEM8A LOC105371184 ATP6V0C PDPK1 RAB11FIP3 CCDC154 FBXL16 PDIA2 CAPN15 LOC107987233 LOC105371045 LOC652276 MIR4516 MCRIP2 GNPTG FAHD1 LOC105371046 RNF151 UBE2I SNORD60 HBA1 CLCN7 DDX11L10 LMF1 MSRB1 MIR3180-5 MIR662 SNORA78 MRPL28 NTHL1 LINC00235 NDUFB10 TMEM204 NARFL CEMP1 FLJ42627 ABCA17P ZNF598 TBL3 PRR35 PRSS29P C16orf59 PKD1 MIR6859-4 RPUSD1 SNHG9 MIR4717 TPSD1 SNRNP25 NPRL3 METTL26 C16orf91 LOC105371047 LOC105371038 PRSS27 MAPK8IP3 MLST8 TBC1D24 IGFALS CACNA1H MIR6767 RHBDL1 SYNGR3 RAB40C MIR3178 BAIAP3 SSTR5-AS1 LMF1-AS1 HN1L METRN MIR5587 HS3ST6 SNHG19 |
nsv471063 | Chr.16:520623 - 852322 on Build GRCh38 | Loss | MIR662 WDR90 LINC00235 NARFL WFIKKN1 JMJD8 PRR25 LOC100287175 MIR3176 PRR35 STUB1 RPUSD1 CCDC78 HAGHL FAM173A METTL26 PIGQ LOC105371038 LOC101929280 LOC105371184 WDR24 RAB11FIP3 RHBDL1 FBXL16 NHLRC4 CHTF18 CAPN15 LOC107987233 RHOT2 RAB40C MCRIP2 GNG13 MSLN METRN MIR5587 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | MIR662 WDR90 LINC00235 NARFL WFIKKN1 JMJD8 PRR25 LOC100287175 MIR3176 PRR35 STUB1 RPUSD1 CCDC78 HAGHL FAM173A METTL26 PIGQ LOC105371038 LOC101929280 LOC105371184 WDR24 RAB11FIP3 RHBDL1 FBXL16 NHLRC4 CHTF18 CAPN15 LOC107987233 RHOT2 RAB40C MCRIP2 GNG13 MSLN METRN MIR5587 |
nsv952898 | Chr.16:495001 - 1132900 on Build GRCh38 | Deletion | MIR662 LOC107984876 WDR90 LINC00235 NARFL SOX8 WFIKKN1 JMJD8 PRR25 LOC100287175 MIR3176 PRR35 STUB1 RPUSD1 SSTR5 CCDC78 HAGHL FAM173A METTL26 PIGQ C1QTNF8 LOC105371038 LOC101929280 LOC105371184 WDR24 RAB11FIP3 RHBDL1 FBXL16 NHLRC4 CHTF18 CAPN15 LOC107987233 RHOT2 RAB40C MCRIP2 GNG13 MSLN SSTR5-AS1 LMF1-AS1 METRN MIR5587 LMF1 |
dgv4823n54 | Chr.16:562698 - 662867 on Build GRCh38 | Loss | LOC107987233 LOC101929280 WDR90 RAB40C MCRIP2 WFIKKN1 METTL26 PIGQ LOC100287175 LOC105371038 PRR35 NHLRC4 |
nsv457304 | Chr.16:554554 - 611142 on Build GRCh38 | Gain | CAPN15 LOC107987233 LOC101929280 RAB40C PIGQ PRR35 NHLRC4 |
nsv1160325 | Chr.16:506925 - 836376 on Build GRCh38 | Duplication | MIR662 WDR90 LINC00235 NARFL WFIKKN1 JMJD8 PRR25 LOC100287175 MIR3176 PRR35 STUB1 RPUSD1 CCDC78 HAGHL FAM173A METTL26 PIGQ LOC105371038 LOC101929280 LOC105371184 WDR24 RAB11FIP3 RHBDL1 FBXL16 NHLRC4 CHTF18 CAPN15 LOC107987233 RHOT2 RAB40C MCRIP2 GNG13 MSLN METRN MIR5587 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |