Assay Details
Target Gene Details
Entrez Gene ID: | 6670 |
Gene Name: | Sp3 transcription factor |
Gene Aliases: |
SPR2 |
Location: |
Chr.2:173906459-173965702 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SP3 | NM_001017371.4 | NP_001017371.3 | ||
NM_001172712.1 | NP_001166183.1 | |||
NM_003111.4 | NP_003102.1 | |||
AB209334.1 | BAD92571.1 | |||
AK304199.1 | ||||
AY070137.1 | AAL58088.1 | |||
AY441957.1 | AAR30505.1 | |||
AY441958.1 | AAR30506.1 | |||
BC042945.2 | AAH42945.2 | |||
BC126414.1 | ||||
BC143928.1 | ||||
CR749323.1 | CAH18178.1 | |||
M97191.2 | AAA36630.2 | |||
X68560.1 | CAA48562.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |