Assay Details
Target Gene Details
Entrez Gene ID: | 3091 |
Gene Name: | hypoxia inducible factor 1 alpha subunit |
Gene Aliases: |
HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78 |
Location: |
Chr.14:61695401-61748259 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A | NM_001243084.1 | 15 | 2981 | NP_001230013.1 |
NM_001530.3 | 15 | 3084 | NP_001521.1 | |
NM_181054.2 | 14 | 2957 | NP_851397.1 | |
AB073325.1 | 14 | 2581 | BAB70608.1 | |
AK299551.1 | 16 | 2786 | ||
AK304315.1 | ||||
AK304436.1 | 14 | 2533 | ||
BC012527.2 | 15 | 2934 | AAH12527.1 | |
BX648795.1 | 15 | 2948 | ||
CN264295.1 | 2 | 371 | ||
FJ790247.1 | 15 | 2982 | ||
U22431.1 | 15 | 2708 | AAC50152.1 | |
U29165.1 | 15 | 2944 | AAC51210.1 | |
X72726.1 | 1 | 282 |
Target Gene Details
Entrez Gene ID: | 100750247 |
Gene Name: | HIF1A antisense RNA 2 |
Gene Aliases: |
3'aHIF-1A, aHIF |
Location: |
Chr.14:61747039-61749089 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A-AS2 | NR_045406.1 | 1 | 1806 | |
BF081731.1 | ||||
BF923330.1 | 1 | 271 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1300 | Chr.14:61737786 - 61782490 on Build GRCh38 | Deletion | HIF1A-AS2 SNAPC1 HIF1A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |