Assay Details
Target Gene Details
Entrez Gene ID: | 8727 |
Gene Name: | catenin alpha like 1 |
Gene Aliases: |
ACRP, CLLP, alpha-CATU |
Location: |
Chr.9:108942569-109013594 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTNNAL1 | NM_001286974.1 | 19 | 2400 | NP_001273903.1 |
NM_003798.3 | 19 | 2363 | NP_003789.1 | |
XM_005252291.3 | 19 | 2325 | XP_005252348.1 | |
XM_017015250.1 | 18 | 2083 | XP_016870739.1 | |
XM_017015251.1 | 18 | 2077 | XP_016870740.1 | |
AB451283.1 | ||||
AB451415.1 | ||||
AF006070.1 | 19 | 2253 | AAC26011.1 | |
AF030233.1 | 19 | 2255 | AAC83460.1 | |
AF080071.1 | 19 | 2250 | AAC69576.1 | |
AF135021.1 | 19 | 2291 | AAF07819.1 | |
AI148526.1 | 1 | 189 | ||
AK022539.1 | 18 | 2045 | ||
AK022834.1 | 19 | 2296 | ||
AK022926.1 | 19 | 2268 | ||
AK123916.1 | 3 | 3194 | ||
AK223230.1 | 19 | 2268 | BAD96950.1 | |
AK314677.1 | ||||
AL050016.1 | 12 | 1075 | CAB43238.1 | |
BC117208.1 | ||||
BC117234.1 | ||||
BU677451.1 | 1 | 211 | ||
U97067.1 | 19 | 2254 | AAC27693.1 |
Target Gene Details
Entrez Gene ID: | 54942 |
Gene Name: | family with sequence similarity 206 member A |
Gene Aliases: |
C9orf6, CG-8, Simiate |
Location: |
Chr.9:108934393-108950745 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM206A | XM_011518816.2 | XP_011517118.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |