Assay Details
Target Gene Details
Entrez Gene ID: | 246175 |
Gene Name: | CCR4-NOT transcription complex subunit 6 like |
Gene Aliases: |
CCR4b |
Location: |
Chr.4:77713387-77820238 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNOT6L | NM_001286790.1 | 12 | 7877 | NP_001273719.1 |
NM_144571.2 | 12 | 7806 | NP_653172.2 | |
XM_011531806.2 | 13 | 7941 | XP_011530108.1 | |
XM_011531807.2 | 13 | 7915 | XP_011530109.1 | |
XM_011531808.2 | 12 | 7887 | XP_011530110.1 | |
XM_011531809.1 | 12 | 7832 | XP_011530111.1 | |
XM_011531810.1 | 12 | 8392 | XP_011530112.1 | |
XM_017007956.1 | 12 | 7829 | XP_016863445.1 | |
XM_017007957.1 | 12 | 8155 | XP_016863446.1 | |
AK124855.1 | 2 | 2470 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829976 | Chr.4:77655503 - 77836644 on Build GRCh38 | Gain | CNOT6L |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |