Assay Details
Target Gene Details
Entrez Gene ID: | 220965 |
Gene Name: | family with sequence similarity 13 member C |
Gene Aliases: |
FAM13C1 |
Location: |
Chr.10:59245933-59363186 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM13C | NM_001001971.2 | 12 | 2061 | NP_001001971.1 |
NM_001143773.1 | 15 | 2526 | NP_001137245.1 | |
NM_001166698.1 | 15 | 2437 | NP_001160170.1 | |
NM_198215.3 | 14 | 2355 | NP_937858.2 | |
XM_005269618.4 | 14 | 2352 | XP_005269675.2 | |
XM_005269619.4 | 12 | 2058 | XP_005269676.2 | |
XM_006717702.3 | 16 | 2356 | XP_006717765.1 | |
XM_006717703.3 | 16 | 2538 | XP_006717766.1 | |
XM_011539490.2 | 15 | 2411 | XP_011537792.1 | |
XM_011539491.2 | 16 | 2457 | XP_011537793.1 | |
XM_017015885.1 | 15 | 2269 | XP_016871374.1 | |
XM_017015886.1 | 15 | 2523 | XP_016871375.1 | |
XM_017015887.1 | 15 | 2273 | XP_016871376.1 | |
XM_017015888.1 | 16 | 2535 | XP_016871377.1 | |
XM_017015889.1 | 16 | 2353 | XP_016871378.1 | |
XM_017015890.1 | 16 | 2592 | XP_016871379.1 | |
XM_017015891.1 | 13 | 2232 | XP_016871380.1 | |
XM_017015892.1 | 13 | 2229 | XP_016871381.1 | |
XM_017015893.1 | 15 | 2333 | XP_016871382.1 | |
AK316542.1 | 15 | 2433 | ||
BC036453.1 | 14 | 2313 | AAH36453.1 | |
BC064431.1 | 12 | 2061 | AAH64431.1 | |
BM714192.1 | 1 | 208 |
Target Gene Details
Entrez Gene ID: | 84457 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein like |
Gene Aliases: |
- |
Location: |
Chr.10:59174811-59247774 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYHIPL | NM_001143774.1 | 5 | 2737 | NP_001137246.1 |
NM_032439.3 | 5 | 2957 | NP_115815.2 | |
XM_011540275.2 | 6 | 3019 | XP_011538577.1 | |
XM_011540276.2 | 5 | 3850 | XP_011538578.1 | |
XM_017016782.1 | 6 | 2838 | XP_016872271.1 | |
XM_017016783.1 | 6 | 3941 | XP_016872272.1 | |
AB058699.1 | 3 | 2298 | BAB47425.1 | |
AL834339.1 | 5 | 2958 | CAD39006.1 | |
BM676577.1 | 1 | 634 | ||
DB551000.1 | 1 | 367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain | FAM13C PHYHIPL |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |