Assay Details
Target Gene Details
Entrez Gene ID: | 6760 |
Gene Name: | SS18, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
SSXT, SYT |
Location: |
Chr.18:26016253-26091217 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SS18 | NM_001007559.2 | 11 | 1914 | NP_001007560.1 |
NM_001308201.1 | 11 | 1889 | NP_001295130.1 | |
NM_005637.3 | 10 | 1821 | NP_005628.2 | |
XM_006722527.2 | 12 | 1940 | XP_006722590.1 | |
XM_011526145.1 | 11 | 1882 | XP_011524447.1 | |
XM_011526147.2 | 13 | 2165 | XP_011524449.1 | |
XM_011526148.2 | 13 | 2152 | XP_011524450.1 | |
XM_011526149.2 | 13 | 2177 | XP_011524451.1 | |
XM_011526150.2 | 12 | 2037 | XP_011524452.1 | |
XM_011526151.2 | 13 | 2166 | XP_011524453.1 | |
XM_011526152.2 | 11 | 1944 | XP_011524454.1 | |
AK296949.1 | 11 | 1872 | ||
X79201.1 | 10 | 1758 | CAA55792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893078 | Chr.18:25391998 - 26039471 on Build GRCh38 | Gain | SS18 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |