Assay Details
Target Gene Details
Entrez Gene ID: | 8309 |
Gene Name: | acyl-CoA oxidase 2 |
Gene Aliases: |
BCOX, BRCACOX, BRCOX, THCCox |
Location: |
Chr.3:58505136-58537202 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOX2 | NM_003500.3 | NP_003491.1 | ||
XM_005265505.1 | XP_005265562.1 | |||
XM_006713340.3 | XP_006713403.1 | |||
XM_017007302.1 | XP_016862791.1 | |||
AK298402.1 | ||||
AK313512.1 | ||||
BC047700.1 | AAH47700.1 | |||
X95190.1 | CAA64489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain | FAM107A FAM3D FHIT C3orf67 ACOX2 C3orf67-AS1 LOC339902 FAM3D-AS1 |
nsv590370 | Chr.3:58472009 - 58557785 on Build GRCh38 | Loss | KCTD6 ACOX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |