Assay Details
Target Gene Details
Entrez Gene ID: | 3558 |
Gene Name: | interleukin 2 |
Gene Aliases: |
IL-2, TCGF, lymphokine |
Location: |
Chr.4:122449479-122456495 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL2 | NM_000586.3 | 4 | 469 | NP_000577.2 |
XM_017008177.1 | XP_016863666.1 | |||
A14844.1 | 4 | 424 | CAA01199.1 | |
AB451325.1 | 4 | 414 | ||
AB451454.1 | 4 | 414 | ||
AF228636.1 | AAG53575.1 | |||
AY283686.1 | 4 | 354 | AAP35033.1 | |
AY523040.1 | 4 | 354 | AAS17753.1 | |
BC066254.1 | 4 | 432 | AAH66254.1 | |
BC066255.1 | 4 | 432 | AAH66255.1 | |
BC066256.1 | 4 | 432 | AAH66256.1 | |
BC066257.1 | 4 | 432 | AAH66257.1 | |
BC070338.1 | 4 | 461 | AAH70338.1 | |
DQ231169.1 | 4 | 354 | ABB20900.1 | |
DQ861285.1 | 4 | 416 | ||
S77834.1 | 4 | 461 | AAD14263.2 | |
S77835.1 | AAD14264.1 | |||
S82692.1 | 4 | 708 | AAB46883.1 | |
U25676.1 | 4 | 470 | AAA70092.1 | |
V00564.1 | 4 | 461 | CAA23827.1 | |
X01586.1 | 4 | 521 | CAA25742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894021 | Chr.4:122306958 - 122473886 on Build GRCh38 | Loss | KIAA1109 ADAD1 IL2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |