Assay Details
Target Gene Details
Entrez Gene ID: | 5562 |
Gene Name: | protein kinase AMP-activated catalytic subunit alpha 1 |
Gene Aliases: |
AMPK, AMPKa1 |
Location: |
Chr.5:40759379-40798195 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKAA1 | NM_006251.5 | 6 | 732 | NP_006242.5 |
NM_206907.3 | 7 | 777 | NP_996790.3 | |
XM_006714482.1 | 5 | 626 | XP_006714545.1 | |
XM_017009624.1 | 7 | 1688 | XP_016865113.1 | |
XM_017009625.1 | 6 | 1633 | XP_016865114.1 | |
AB022017.1 | 6 | 722 | BAA36547.1 | |
AF100763.1 | 6 | 702 | AAD43027.1 | |
AK307546.1 | 7 | 759 | ||
AK312947.1 | 6 | 714 | ||
BC037303.2 | 6 | 714 | AAH37303.1 | |
BC048980.1 | 7 | 956 | AAH48980.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |