Assay Details
Target Gene Details
Entrez Gene ID: | 130916 |
Gene Name: | mitochondrial transcription termination factor 4 |
Gene Aliases: |
MTERFD2 |
Location: |
Chr.2:241072168-241103367 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTERF4 | NR_028049.1 | 7 | 4277 | |
NR_028050.1 | 5 | 2469 | ||
NR_028051.1 | 4 | 2284 | ||
XM_006712294.1 | 5 | 2738 | XP_006712357.1 | |
XM_006712295.3 | 5 | 2894 | XP_006712358.1 | |
XM_011510634.2 | 7 | 4580 | XP_011508936.1 | |
XM_017003379.1 | 7 | 5208 | XP_016858868.1 | |
XM_017003380.1 | XP_016858869.1 | |||
XM_017003381.1 | 8 | 5044 | XP_016858870.1 | |
XM_017003382.1 | 8 | 4557 | XP_016858871.1 | |
XM_017003383.1 | 8 | 5330 | XP_016858872.1 | |
XM_017003384.1 | 8 | 5305 | XP_016858873.1 | |
XM_017003385.1 | XP_016858874.1 | |||
XM_017003386.1 | 7 | 5679 | XP_016858875.1 | |
XM_017003387.1 | 6 | 5231 | XP_016858876.1 | |
XM_017003389.1 | 6 | 3991 | XP_016858878.1 | |
XM_017003390.1 | 5 | 4464 | XP_016858879.1 | |
XM_017003391.1 | 5 | 5484 | XP_016858880.1 | |
AK025374.1 | 1 | 1529 | ||
AK055474.1 | 4 | 3674 | ||
AK093695.1 | 5 | 2424 | ||
AK124444.1 | ||||
BX648184.1 | 7 | 4243 |
Target Gene Details
Entrez Gene ID: | 25992 |
Gene Name: | sushi, nidogen and EGF like domains 1 |
Gene Aliases: |
SST3, Snep |
Location: |
Chr.2:240997409-241095568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SNED1 | NM_001080437.1 | NP_001073906.1 | ||
XM_011510931.2 | XP_011509233.1 | |||
XM_011510932.2 | XP_011509234.1 | |||
XM_017003773.1 | XP_016859262.1 | |||
XM_017003774.1 | XP_016859263.1 | |||
XM_017003775.1 | XP_016859264.1 | |||
XM_017003776.1 | XP_016859265.1 | |||
XM_017003777.1 | XP_016859266.1 | |||
XM_017003778.1 | XP_016859267.1 | |||
XM_017003779.1 | XP_016859268.1 | |||
XM_017003780.1 | XP_016859269.1 | |||
XM_017003781.1 | XP_016859270.1 | |||
XM_017003782.1 | XP_016859271.1 | |||
AF439717.1 | AAQ04558.1 | |||
AK074062.1 | BAB84888.1 | |||
AK074075.1 | BAB84901.1 | |||
BC027939.1 | AAH27939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575345 | Chr.2:241086207 - 241153068 on Build GRCh38 | Gain | MTERF4 PASK PPP1R7 SNED1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |