Assay Details
Target Gene Details
Entrez Gene ID: | 170591 |
Gene Name: | S100 calcium binding protein Z |
Gene Aliases: |
Gm625, S100-zeta |
Location: |
Chr.5:76849953-76952882 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100Z | NM_130772.3 | 4 | 535 | NP_570128.2 |
XM_011543239.2 | 4 | 583 | XP_011541541.1 | |
XM_011543240.2 | 4 | 583 | XP_011541542.1 | |
XM_011543241.2 | 4 | 583 | XP_011541543.1 | |
XM_011543244.2 | 4 | 550 | XP_011541546.1 | |
XM_011543245.2 | 3 | 1320 | XP_011541547.1 | |
XM_017009171.1 | 4 | 775 | XP_016864660.1 | |
XM_017009172.1 | 4 | 786 | XP_016864661.1 | |
XM_017009173.1 | 3 | 464 | XP_016864662.1 | |
AF437876.1 | 2 | 275 | AAL30893.1 | |
BC022320.1 | 4 | 536 | AAH22320.1 | |
BX281742.1 | 4 | 437 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524024 | Chr.5:76870193 - 76891952 on Build GRCh38 | Loss | S100Z |
esv34133 | Chr.5:76842871 - 76943505 on Build GRCh38 | Loss | S100Z |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |