Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001170695.1 | NP_001164166.1 | ||
NM_001170696.1 | NP_001164167.1 | |||
NM_001170697.1 | NP_001164168.1 | |||
NM_001170698.1 | NP_001164169.1 | |||
NM_001170699.1 | NP_001164170.1 | |||
NM_001321336.1 | NP_001308265.1 | |||
NM_001321337.1 | NP_001308266.1 | |||
NM_032598.4 | NP_115987.2 | |||
AF367472.1 | AAK53408.1 | |||
AK295299.1 | ||||
AK301892.1 | ||||
AY032684.1 | AAK51120.1 | |||
AY035867.1 | AAK61373.1 | |||
AY035868.1 | AAK61374.1 | |||
BC029483.1 | AAH29483.1 | |||
BG716998.1 | ||||
BI463957.1 | ||||
BP371122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR3A3 OR1D4 TRPV3 ASPA TRPV1 OR3A1 OR1E1 OR3A4P SPATA22 OR1E2 OR1A1 OR1A2 OR3A2 |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | MIR6776 P2RX5 OR1G1 OR1D4 TRPV3 CTNS CCDC92B MIR1253 SPATA22 LOC105371592 P2RX5-TAX1BP3 PAFAH1B1 OR1D2 OR1A2 LOC101927911 SHPK CAMKK1 RAP1GAP2 OR3A3 EMC6 ATP2A3 LOC105371591 P2RX1 NCBP3 ASPA TRPV1 OR3A1 CLUH OR1E1 OR3A4P OR1E2 ITGAE OR1A1 GSG2 TAX1BP3 OR1D5 OR3A2 |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR3A1 OR1E1 OR3A4P SPATA22 OR1E2 OR3A3 OR3A2 ASPA |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | MIR6776 RAP1GAP2 OR3A3 OR1G1 LOC105371591 OR1D4 TRPV3 ASPA OR3A1 CCDC92B CLUH OR1E1 OR3A4P MIR1253 SPATA22 LOC105371592 OR1E2 PAFAH1B1 OR1A1 OR1D2 OR1A2 LOC101927911 OR1D5 OR3A2 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | CAMKK1 OR3A3 P2RX5 EMC6 NCBP3 TRPV3 CTNS ASPA TRPV1 OR1E1 SPATA22 P2RX5-TAX1BP3 OR1E2 ITGAE GSG2 TAX1BP3 SHPK OR3A2 |
nsv528368 | Chr.17:3459775 - 3507608 on Build GRCh38 | Gain | SPATA22 ASPA |
nsv521277 | Chr.17:3390044 - 3468885 on Build GRCh38 | Gain | OR1E1 SPATA22 OR1E2 OR3A3 |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR1E1 SPATA22 OR1E2 OR3A3 TRPV3 OR3A2 ASPA |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | TRPV1 SPATA22 OR1E2 OR3A3 TRPV3 SHPK ASPA |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |