Assay Details
Target Gene Details
Entrez Gene ID: | 7433 |
Gene Name: | vasoactive intestinal peptide receptor 1 |
Gene Aliases: |
HVR1, II, PACAP-R-2, PACAP-R2, RDC1, V1RG, VAPC1, VIP-R-1, VIPR, VIRG, VPAC1, VPAC1R, VPCAP1R |
Location: |
Chr.3:42489299-42537573 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VIPR1 | NM_001251882.1 | NP_001238811.1 | ||
NM_001251883.1 | NP_001238812.1 | |||
NM_001251884.1 | NP_001238813.1 | |||
NM_001251885.1 | NP_001238814.1 | |||
NM_004624.3 | NP_004615.2 | |||
XM_005265437.2 | XP_005265494.1 | |||
XM_005265438.3 | XP_005265495.1 | |||
XM_005265439.2 | XP_005265496.1 | |||
XM_011534079.1 | XP_011532381.1 | |||
XM_011534080.2 | XP_011532382.1 | |||
AK056819.1 | ||||
AK293548.1 | ||||
AK294609.1 | ||||
AK298115.1 | ||||
AK314334.1 | ||||
BC064424.1 | AAH64424.1 | |||
DA200439.1 | ||||
EF577396.1 | ||||
L13288.1 | AAA36805.1 | |||
L20295.1 | AAA36802.1 | |||
X75299.1 | CAA53046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527430 | Chr.3:42492550 - 42515707 on Build GRCh38 | Loss | VIPR1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |