Assay Details
Target Gene Details
Entrez Gene ID: | 3579 |
Gene Name: | C-X-C motif chemokine receptor 2 |
Gene Aliases: |
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB |
Location: |
Chr.2:218125290-218137253 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCR2 | NM_001168298.1 | 4 | 279 | NP_001161770.1 |
NM_001557.3 | 3 | 487 | NP_001548.1 | |
XM_005246530.3 | 4 | 259 | XP_005246587.1 | |
XM_017003990.1 | 4 | 239 | XP_016859479.1 | |
XM_017003991.1 | 2 | 150 | XP_016859480.1 | |
XM_017003992.1 | 2 | 420 | XP_016859481.1 | |
AK290906.1 | 3 | 385 | ||
AK312664.1 | 3 | 378 | ||
BC037961.1 | 3 | 441 | AAH37961.2 | |
DA670033.1 | 3 | 487 | ||
DA674925.1 | 4 | 279 | ||
L19593.1 | 3 | 477 | AAB59437.1 | |
M73969.1 | 1 | 94 | AAA83148.1 | |
M94582.1 | 2 | 121 | AAA36108.1 | |
U11872.1 | AAA64380.1 | |||
U11873.1 | AAA64381.1 | |||
U11874.1 | AAA64382.1 | |||
U11875.1 | AAA64383.1 | |||
U11876.1 | AAA64384.1 | |||
U11877.1 | AAA64385.1 | |||
U11878.1 | AAA64386.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |