Assay Details
Target Gene Details
Entrez Gene ID: | 113622 |
Gene Name: | ADP-ribosylhydrolase like 1 |
Gene Aliases: |
ARH2 |
Location: |
Chr.13:113421940-113453524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADPRHL1 | NM_138430.4 | NP_612439.2 | ||
NM_199162.2 | NP_954631.1 | |||
AJ313429.1 | CAC86114.1 | |||
BC009755.2 | AAH09755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain | PCID2 DCUN1D2 PROZ GRTP1 MCF2L TMCO3 MIR8075 F10-AS1 LOC101928841 LOC105370378 ADPRHL1 F10 GRTP1-AS1 CUL4A LAMP1 F7 LOC102724474 TFDP1 |
dgv3449n54 | Chr.13:113433286 - 113436681 on Build GRCh38 | Loss | ADPRHL1 |
esv3633609 | Chr.13:113431952 - 113440673 on Build GRCh38 | Loss | ADPRHL1 |
dgv1744n100 | Chr.13:113384831 - 113442576 on Build GRCh38 | Gain | ADPRHL1 LOC101928841 |
esv2677394 | Chr.13:113431938 - 113440736 on Build GRCh38 | Deletion | ADPRHL1 |
nsv951928 | Chr.13:113392386 - 113435785 on Build GRCh38 | Deletion | ADPRHL1 LOC101928841 |
nsv563321 | Chr.13:113369086 - 113452383 on Build GRCh38 | Loss | ADPRHL1 LOC101928841 |
esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss | DCUN1D2 ADPRHL1 TMCO3 LOC101928841 |
nsv1044941 | Chr.13:113369130 - 113433835 on Build GRCh38 | Gain | ADPRHL1 LOC101928841 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |