Assay Details
Target Gene Details
Entrez Gene ID: | 1796 |
Gene Name: | docking protein 1 |
Gene Aliases: |
P62DOK, pp62 |
Location: |
Chr.2:74549020-74557551 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOK1 | NM_001197260.1 | NP_001184189.1 | ||
NM_001318866.1 | NP_001305795.1 | |||
NM_001318867.1 | NP_001305796.1 | |||
NM_001318868.1 | NP_001305797.1 | |||
NM_001318869.1 | NP_001305798.1 | |||
NM_001381.4 | NP_001372.1 | |||
XM_017003528.1 | XP_016859017.1 | |||
AK055944.1 | ||||
AK127127.1 | ||||
AK127656.1 | ||||
AK222883.1 | BAD96603.1 | |||
AK300097.1 | ||||
BC111055.1 | AAI11056.1 | |||
BQ575925.1 |
Target Gene Details
Entrez Gene ID: | 130951 |
Gene Name: | meiosis 1 associated protein |
Gene Aliases: |
C2orf65, D6Mm5e, SPATA37 |
Location: |
Chr.2:74557640-74648338 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
M1AP | XM_005264152.3 | XP_005264209.1 | ||
XM_006711946.3 | XP_006712009.1 | |||
XM_011532548.2 | XP_011530850.1 | |||
XM_011532549.2 | XP_011530851.1 | |||
XM_011532550.2 | XP_011530852.1 | |||
XM_011532551.2 | XP_011530853.1 | |||
XM_011532552.2 | XP_011530854.1 | |||
XM_011532554.2 | XP_011530856.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |