Assay Details
Target Gene Details
Entrez Gene ID: | 347 |
Gene Name: | apolipoprotein D |
Gene Aliases: |
- |
Location: |
Chr.3:195568702-195584205 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOD | NM_001647.3 | 1 | 259 | NP_001638.1 |
AK294523.1 | 1 | 75 | ||
AK312090.1 | ||||
BC007402.2 | AAH07402.1 | |||
BU195270.1 | 1 | 172 | ||
DA892855.1 | 1 | 259 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5016n100 | Chr.3:195552380 - 195720039 on Build GRCh38 | Gain+Loss | SDHAP2 LOC105374297 LINC00969 APOD MIR570 |
nsv1001387 | Chr.3:195465410 - 195934667 on Build GRCh38 | Gain | SDHAP2 MUC20 LOC101929697 LOC105374297 MIR6829 LINC00969 PPP1R2 APOD MIR570 LOC105374299 TNK2 MUC4 |
dgv5015n100 | Chr.3:195532487 - 195751823 on Build GRCh38 | Loss | SDHAP2 MUC20 LOC105374297 LINC00969 PPP1R2 APOD MIR570 MUC4 |
nsv1006229 | Chr.3:195350692 - 195747880 on Build GRCh38 | Gain | SDHAP2 MUC20 LOC105374297 LINC00969 ACAP2 PPP1R2 APOD MIR570 MUC4 |
nsv428430 | Chr.3:195478549 - 196102555 on Build GRCh38 | Gain+Loss | MUC20 LOC101929697 TFRC SDHAP1 MIR570 LOC105374299 TNK2 SDHAP2 LOC105374297 MIR6829 LINC00969 PPP1R2 APOD LOC105374301 MUC4 |
esv2763776 | Chr.3:195514028 - 195775248 on Build GRCh38 | Gain+Loss | SDHAP2 MUC20 LOC105374297 LINC00969 PPP1R2 APOD MIR570 MUC4 |
dgv5014n100 | Chr.3:195514016 - 195747880 on Build GRCh38 | Gain | SDHAP2 MUC20 LOC105374297 LINC00969 PPP1R2 APOD MIR570 MUC4 |
esv2759213 | Chr.3:195382058 - 196344722 on Build GRCh38 | Gain+Loss | MUC20 LOC101929697 TFRC SDHAP1 PCYT1A MIR570 LOC105374299 TNK2 TCTEX1D2 SDHAP2 SLC51A LOC105374297 TM4SF19-TCTEX1D2 MIR6829 LINC00969 LINC00885 ACAP2 PPP1R2 APOD LOC105374301 TM4SF19-AS1 ZDHHC19 TM4SF19 MUC4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |