Assay Details
Target Gene Details
Entrez Gene ID: | 27230 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
RAMP4 |
Location: |
Chr.3:150541993-150603177 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERP1 | NM_014445.3 | 3 | 1732 | NP_055260.1 |
XM_011512698.2 | 4 | 2397 | XP_011511000.1 | |
AK054644.1 | 1 | 558 | ||
AK090933.1 | 4 | 1544 | ||
AK093929.1 | 4 | 1553 | ||
AK098135.1 | 4 | 1448 | ||
AK125413.1 | 4 | 2498 | ||
AL136807.1 | 3 | 1541 | CAB66741.1 | |
BC108314.1 | 3 | 1580 | AAI08315.1 | |
BC112364.1 | 3 | 1539 | AAI12365.1 | |
BC112365.1 | 3 | 1539 | AAI12366.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |