Assay Details
Target Gene Details
Entrez Gene ID: | 2734 |
Gene Name: | golgi glycoprotein 1 |
Gene Aliases: |
CFR-1, ESL-1, MG-160, MG160 |
Location: |
Chr.16:74447427-74607144 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLG1 | NM_001145666.1 | NP_001139138.1 | ||
NM_001145667.1 | NP_001139139.1 | |||
NM_012201.5 | NP_036333.2 | |||
NR_027264.1 | ||||
NR_027265.1 | ||||
AK025457.1 | ||||
AK026972.1 | ||||
AK027032.1 | ||||
AK131501.1 | 4 | 1890 | ||
AK172806.1 | BAD18777.1 | |||
AK304156.1 | ||||
BC060822.1 | AAH60822.1 | |||
U28811.1 | AAB02178.1 | |||
U64791.1 | AAB06460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833279 | Chr.16:74456848 - 74629175 on Build GRCh38 | Loss |
![]() ![]() |
nsv833278 | Chr.16:74382046 - 74550225 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2762054 | Chr.16:74303138 - 74473158 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833277 | Chr.16:74300889 - 74464235 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1065964 | Chr.16:74054192 - 74541706 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639067 | Chr.16:74445112 - 74488116 on Build GRCh38 | Gain |
![]() |
esv3571587 | Chr.16:74432668 - 74469268 on Build GRCh38 | Gain |
![]() |
nsv509632 | Chr.16:74311644 - 74471617 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2758656 | Chr.16:74300882 - 74553287 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)