Assay Details
Target Gene Details
Entrez Gene ID: | 391253 |
Gene Name: | serine peptidase inhibitor, Kunitz type 4 |
Gene Aliases: |
C20orf137, SPINT3, dJ601O1.1 |
Location: |
Chr.20:45722349-45725696 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINT4 | NM_178455.1 | NP_848550.1 | ||
AY372173.1 | AAR17082.1 | |||
AY372174.1 | AAR17083.1 |
Target Gene Details
Entrez Gene ID: | 140686 |
Gene Name: | WAP four-disulfide core domain 3 |
Gene Aliases: |
WAP14, dJ447F3.3 |
Location: |
Chr.20:45708860-45791908 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC3 | XM_017027668.1 | XP_016883157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv23805 | Chr.20:45724172 - 45743377 on Build GRCh38 | Gain |
![]() ![]() |
esv2751916 | Chr.20:45722292 - 45771812 on Build GRCh38 | Loss |
![]() ![]() |
dgv7591n54 | Chr.20:45722456 - 45734110 on Build GRCh38 | Loss |
![]() ![]() |
esv33602 | Chr.20:45722083 - 45750234 on Build GRCh38 | Loss |
![]() ![]() |
dgv7590n54 | Chr.20:45709047 - 45749938 on Build GRCh38 | Loss |
![]() ![]() |
dgv4320n100 | Chr.20:45713700 - 45758128 on Build GRCh38 | Loss |
![]() ![]() |
esv2678145 | Chr.20:45720211 - 45750898 on Build GRCh38 | Deletion |
![]() ![]() |
nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3557119 | Chr.20:45720144 - 45750978 on Build GRCh38 | Deletion |
![]() ![]() |
esv2763671 | Chr.20:45719923 - 45752208 on Build GRCh38 | Loss |
![]() ![]() |
nsv586116 | Chr.20:45714903 - 45734110 on Build GRCh38 | Loss |
![]() ![]() |
esv3645939 | Chr.20:45719986 - 45751040 on Build GRCh38 | Loss |
![]() ![]() |
dgv1247e212 | Chr.20:45708247 - 45752976 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2751915 | Chr.20:45708077 - 45749572 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv4321n100 | Chr.20:45719911 - 45754523 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)