Assay Details
Target Gene Details
Entrez Gene ID: | 137902 |
Gene Name: | peroxidasin like |
Gene Aliases: |
PMR1, PRM1, VPO2 |
Location: |
Chr.8:51319575-51811031 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PXDNL | NM_144651.4 | 23 | 4507 | NP_653252.3 |
XM_005251168.3 | 9 | 2654 | XP_005251225.1 | |
XM_006716420.3 | 21 | 5805 | XP_006716483.1 | |
XM_011517456.2 | 22 | 6021 | XP_011515758.1 | |
XM_011517457.2 | 22 | 6021 | XP_011515759.1 | |
XM_011517458.2 | 21 | 4181 | XP_011515760.1 | |
XM_017013041.1 | 20 | 4142 | XP_016868530.1 | |
AI127081.1 | 1 | 306 | ||
AK058200.1 | 7 | 1727 | BAB71713.1 | |
AK131524.1 | 5 | 1959 | BAD18663.1 | |
AY877349.1 | 23 | 4507 | ||
DB337228.1 | 1 | 221 | ||
EU170240.1 | ||||
KJ545488.1 | 19 | 3972 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |