Assay Details
Target Gene Details
Entrez Gene ID: | 4715 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B9 |
Gene Aliases: |
B22, CI-B22, LYRM3, UQOR22 |
Location: |
Chr.8:124539102-124549986 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFB9 | NM_001278645.1 | NP_001265574.1 | ||
NM_001278646.1 | NP_001265575.1 | |||
NM_001311168.1 | NP_001298097.1 | |||
NM_005005.2 | NP_004996.1 | |||
AF044956.1 | AAD42057.1 | |||
AF067168.1 | AAD32452.1 | |||
AF261090.1 | AAF99683.1 | |||
AI907018.1 | ||||
AK026538.1 | ||||
AK302286.1 | ||||
AK313432.1 | ||||
AL136714.1 | CAI46222.1 | |||
BC007672.2 | AAH07672.1 | |||
BG703246.1 | ||||
BQ685604.1 | ||||
CN410734.1 | ||||
DB504123.1 | ||||
S82655.1 | AAB46797.2 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |