Assay Details
Target Gene Details
Entrez Gene ID: | 57575 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
OL-PCDH, PCDH19 |
Location: |
Chr.4:133149290-133208606 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCDH10 | NM_020815.2 | 1 | 3262 | NP_065866.1 |
NM_032961.2 | 1 | 3262 | NP_116586.1 | |
XM_011532150.1 | 1 | 3262 | XP_011530452.1 | |
AB037821.1 | 1 | 3237 | ||
AK024636.1 | 1 | 526 | BAB14940.1 | |
AK314377.1 | 1 | 2612 | ||
AL157470.1 | 1 | 319 | CAB75667.1 | |
AY013874.1 | 1 | 3237 | AAK21987.1 | |
BC111560.1 | 1 | 2600 | ||
KJ534913.1 | 1 | 2411 | ||
KJ534914.1 | 2 | 2243 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |