Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK6 | NM_001012964.2 | 5 | 975 | NP_001012982.1 |
NM_001012965.2 | 4 | 927 | NP_001012983.1 | |
NM_001319948.1 | 5 | 621 | NP_001306877.1 | |
NM_001319949.1 | 4 | 818 | NP_001306878.1 | |
NM_002774.3 | 6 | 778 | NP_002765.1 | |
AF013988.1 | 6 | 680 | AAB66483.1 | |
AK314897.1 | 6 | 683 | ||
AY279383.1 | 5 | 622 | ||
AY318867.1 | 5 | 784 | AAP82446.1 | |
AY318868.1 | 4 | 613 | ||
AY318869.1 | 5 | 770 | AAP82448.1 | |
AY318870.1 | 4 | 722 | ||
AY457039.1 | AAR23148.1 | |||
BC015525.2 | 5 | 751 | AAH15525.1 | |
BT006852.1 | 4 | 534 | AAP35498.1 | |
D78203.1 | 6 | 689 | BAA11306.1 | |
DQ223012.1 | 4 | 626 | ABB04464.1 | |
U62801.1 | 6 | 779 | AAB07113.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |