Assay Details
Target Gene Details
Entrez Gene ID: | 10516 |
Gene Name: | fibulin 5 |
Gene Aliases: |
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50 |
Location: |
Chr.14:91869411-91947702 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBLN5 | NM_006329.3 | 11 | 2311 | NP_006320.2 |
XM_005267267.3 | 11 | 2146 | XP_005267324.1 | |
XM_011536356.1 | 12 | 2325 | XP_011534658.1 | |
XM_011536357.1 | 12 | 2480 | XP_011534659.1 | |
XM_011536358.1 | 13 | 2580 | XP_011534660.1 | |
XM_017020929.1 | 12 | 2401 | XP_016876418.1 | |
AA812956.1 | 1 | 327 | ||
AF093118.1 | AAC62107.1 | |||
AF112152.1 | 11 | 2006 | AAD41768.1 | |
AJ133490.1 | 11 | 1838 | CAB38568.1 | |
AK074540.1 | 5 | 1217 | ||
AK075147.1 | 11 | 2043 | ||
AK094032.1 | 1 | 2001 | ||
AK125600.1 | 2 | 1973 | ||
AL110148.1 | 1 | 310 | ||
AY358898.1 | 11 | 2287 | AAQ89257.1 | |
BC022280.1 | 11 | 2021 | AAH22280.1 | |
BX248290.1 | 12 | 2131 | CAD62618.1 | |
BX537531.1 | 1 | 6335 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |